missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15498015
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15498015 250 mg 250mg
15454887 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15498015

Brand: Thermo Scientific Alfa Aesar H32201.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Fluoro-2-(trifluoromethoxy)benzyl bromide, is used as a primary and secondary intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092460-88-2
Molecular Formula C8H5BrF4O
Molecular Weight (g/mol) 273.025
MDL Number MFCD11519337
InChI Key VUUGICQPCOWAIC-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy-5-fluorobenzyl bromide, 5-fluoro-2-trifluoromethoxy benzyl bromide, 2-bromomethyl-4-fluoro-1-trifluoromethoxy benzene
PubChem CID 53421164
IUPAC Name 2-(bromomethyl)-4-fluoro-1-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1F)CBr)OC(F)(F)F

Specifications

Refractive Index 1.466
Quantity 250 mg
UN Number UN3265
Formula Weight 273.02
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.