missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrotoluene, 98+%

Product Code. 11451678
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451678 10 g 10g
11461678 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451678 Supplier Thermo Scientific Alfa Aesar Supplier No. A13944.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 446-33-3 | C7H6FNO2 | 155.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-33-3
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007283
InChI Key JHFOWEGCZWLHNW-UHFFFAOYSA-N
Synonym 5-fluoro-2-nitrotoluene, 3-fluoro-6-nitrotoluene, 4-fluoro-2-methylnitrobenzene, 2-nitro-5-fluoro toluene, benzene, 4-fluoro-2-methyl-1-nitro, toluene, 5-fluoro-2-nitro, 2-nitro-5-fluorotoluene, 2-methyl-4-fluoronitrobenzene, 3-methyl-4-nitrofluorobenzene, 4-fluoro-2-methyl-1-nitro-benzene
PubChem CID 94956
IUPAC Name 4-fluoro-2-methyl-1-nitrobenzene
SMILES CC1=C(C=CC(=C1)F)[N+](=O)[O-]

Specifications

Melting Point ∼20°C
Density 1.27
Boiling Point 97°C to 98°C (10 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.528
Quantity 10 g
UN Number UN2810
Beilstein 2046652
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2-nitrotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.