missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 25 g |
Formula Weight | 167.21 |
Percent Purity | 97% |
Chemical Name or Material | 5-Fluoro-2-methylphenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'