missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylphenyl isothiocyanate, 97%, Maybridge™

Brand:  Maybridge SB01782DE

Code : 31

Additional Details : CAS Number : 175205-39-7 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 10046654

  • £24.11 / 5g

Estimated Shipment: 20-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

175205-39-7
167.201
5-fluoro-2-methylphenyl isothiocyanate, 2-methyl-5-fluorophenyl isothiocyanate, 5-fluoro-o-tolyl isothiocyanate, 5-fluoro-2-methylphenylisothiocyanate, isothiocyanic acid 5-fluoro-o-tolyl ester, acmc-20am3d, 4-fluoro-2-isothiocyanatotoluene, 2-methyl-5-fluorophenylisothiocyanate, 5-fluoro-2-methylbenzenisothiocyanate, 5-fluoro-2-methylphenyl-isothiocyanate
4-fluoro-2-isothiocyanato-1-methylbenzene
C8H6FNS
QVCYTKXGZLOVFA-UHFFFAOYSA-N
519443
CC1=C(C=C(C=C1)F)N=C=S

Specifications

5-Fluoro-2-methylphenyl isothiocyanate
5g
97%
5-fluoro-2-methylphenyl isothiocyanate, 2-methyl-5-fluorophenyl isothiocyanate, 5-fluoro-o-tolyl isothiocyanate, 5-fluoro-2-methylphenylisothiocyanate, isothiocyanic acid 5-fluoro-o-tolyl ester, acmc-20am3d, 4-fluoro-2-isothiocyanatotoluene, 2-methyl-5-fluorophenylisothiocyanate, 5-fluoro-2-methylbenzenisothiocyanate, 5-fluoro-2-methylphenyl-isothiocyanate
CC1=C(C=C(C=C1)F)N=C=S
167.201
167.21
Amber glass bottle
175205-39-7
C8H6FNS
QVCYTKXGZLOVFA-UHFFFAOYSA-N
4-fluoro-2-isothiocyanato-1-methylbenzene
519443
97%
Videos
SDS Documents
Certificates