missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzoyl chloride, 98%

Product Code. 11353217
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353217 5 g 5g
11343217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353217 Supplier Thermo Scientific Alfa Aesar Supplier No. L13750.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21900-39-0 | C8H6ClFO | 172.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21900-39-0
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.58
MDL Number MFCD00236681
InChI Key HBAQRSDZSVESSF-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylbenzoylchloride, 5-fluoro-2-methyl-benzoyl chloride, benzoyl chloride,5-fluoro-2-methyl, acmc-1ciaq, 2-methyl-5-fluorobenzoyl chloride, 5-fluoro-2-methyl benzoylchloride, 5-fluoro-2-methyl benzoyl chloride, benzoyl chloride, 5-fluoro-2-methyl, 5-fluoro-2-methylbenzoyl chloride
PubChem CID 2737383
IUPAC Name 5-fluoro-2-methylbenzoyl chloride
SMILES CC1=CC=C(F)C=C1C(Cl)=O

Specifications

Density 1.27
Boiling Point 108°C (24 mmHg)
Refractive Index 1.532
Quantity 5 g
UN Number UN3265
Beilstein 2574820
Sensitivity Moisture sensitive
Formula Weight 172.59
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methylbenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.