missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzenesulfonyl chloride, 97%

Product Code. 11490483
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490483 1 g 1g
11400493 5 g 5g
11410493 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490483 Supplier Thermo Scientific Alfa Aesar Supplier No. A12641.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 445-05-6 | C7H6ClFO2S | 208.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-05-6
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.63
MDL Number MFCD00052958
InChI Key NPEWBMJAUUSBLE-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylbenzene-1-sulfonyl chloride, 5-fluoro-2-methylbenzenesulphonyl chloride, 5-fluoro-2-methylbenzenesulfonylchloride, 5-fluoro-2-methyl-benzenesulfonyl chloride, chloro 5-fluoro-2-methylphenyl sulfone, benzenesulfonyl chloride, 5-fluoro-2-methyl, 2-methyl-5-fluorobenzenesulfonyl chloride, pubchem5165, acmc-20ai7v, 5-fluoro-2-methylphenylsulfonyl chloride
PubChem CID 136286
IUPAC Name 5-fluoro-2-methylbenzenesulfonyl chloride
SMILES CC1=CC=C(F)C=C1S(Cl)(=O)=O

Specifications

Density 1.43
Boiling Point 130°C to 132°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 1 g
UN Number UN3265
Beilstein 2643541
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.