missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10226004
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10226004

Brand: Thermo Scientific Maybridge TL00317EB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 445-05-6
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.63
MDL Number MFCD00052958
InChI Key NPEWBMJAUUSBLE-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylbenzene-1-sulfonyl chloride, 5-fluoro-2-methylbenzenesulphonyl chloride, 5-fluoro-2-methylbenzenesulfonylchloride, 5-fluoro-2-methyl-benzenesulfonyl chloride, chloro 5-fluoro-2-methylphenyl sulfone, benzenesulfonyl chloride, 5-fluoro-2-methyl, 2-methyl-5-fluorobenzenesulfonyl chloride, pubchem5165, acmc-20ai7v, 5-fluoro-2-methylphenylsulfonyl chloride
PubChem CID 136286
SMILES CC1=CC=C(F)C=C1S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula F(CH3)C6H3SO2Cl
Quantity 25 g
IUPAC Name 5-fluoro-2-methylbenzenesulfonyl chloride
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methylbenzene-1-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.