missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methyl-1H-indene-3-acetic acid, 97%, Thermo Scientific™

Product Code. 15446496
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446496 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446496 Supplier Thermo Scientific Alfa Aesar Supplier No. H61410.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32004-66-3
Molecular Formula C12H11FO2
MDL Number MFCD01076194
Quantity 250 mg
Synonym 5-fluoro-2-methyl-1h-inden-3-yl acetic acid, 2-5-fluoro-2-methyl-1h-inden-3-yl acetic acid, 5-fluoro-2-methyl-1h-indene-3-acetic acid, 5-fluoro-2-methylindene-3-acetic acid, 5-fluoro-2-methyl-3-indenyl-acetic acid, 6-fluoro-2-methyl-3h-inden-1-yl acetic acid, 2-6-fluoro-2-methyl-3h-inden-1-yl acetic acid, fluoromethylindenylaceticacid, 5-fluoro-2-methyl-3-indenylacetic acid, 5-fluoro-2-methylinden-3-yl acetic acid
InChI Key QDDPPRDVFIJASZ-UHFFFAOYSA-N
SMILES CC1=C(C2=C(C1)C=CC(=C2)F)CC(=O)O
IUPAC Name 2-(6-fluoro-2-methyl-3H-inden-1-yl)acetic acid
Molecular Weight (g/mol) 206.216
PubChem CID 3015608
Formula Weight 206.21
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-methyl-1H-indene-3-acetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.