missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxyphenylboronic acid, 98%, Thermo Scientific™

Product Code. 10881671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10881671 1 g Glass bottle 1g
10702962 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10881671 Supplier Thermo Scientific Acros Supplier No. 393890010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Identificadores de productos químicos

CAS 179897-94-0
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD01863526
InChI Key CCQKIRUMTHHPSX-UHFFFAOYSA-N
Synonym 5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl
PubChem CID 2782673
IUPAC Name (5-fluoro-2-methoxyphenyl)boronic acid
SMILES COC1=CC=C(F)C=C1B(O)O

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point 156.0°C to 158.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 169.95
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-methoxyphenylboronic acid
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 5-Fluoro-2-methoxyphenylboronic acid
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.