missing translation for 'onlineSavingsMsg'
Learn More

5'-Fluoro-2'-hydroxyacetophenone, 97%, ACROS Organics™

Product Code. 10760955 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10760955 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10760955 Supplier Thermo Scientific Acros Supplier No. 277930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 394-32-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 54°C to 59°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H7FO2
Linear Formula FC6H3(OH)COCH3
MDL Number MFCD00011668
Quantity 5 g
Synonym 5'-fluoro-2'-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl ethanone, 1-5-fluoro-2-hydroxyphenyl ethan-1-one, 5-fluoro-2-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl-1-ethanone, 2-hydroxy-5-fluoroacetophenone, 2'-hydroxy-5'-fluoroacetophenone, ethanone, 1-5-fluoro-2-hydroxyphenyl, 5'-fluoro-2'-hydroxy acetophenone, 5'-fluoro-2'-hydroxy-acetophenone
InChI Key KOFFXZYMDLWRHX-UHFFFAOYSA-N
SMILES CC(=O)C1=C(C=CC(=C1)F)O
IUPAC Name 1-(5-fluoro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol) 154.14
PubChem CID 95992
Formula Weight 154.14
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5'-Fluoro-2'-hydroxyacetophenone, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.