missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2'-deoxyuridine, 98+%

Product Code. p-7046834
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11389205

Brand: Thermo Scientific Alfa Aesar L16497.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol

5-Fluoro-2'-deoxyuridine acts as an antiviral and antineoplastic agent. It is also employed for renal function diagnosis. Further, it is used as an experimental anticancer agent, which shows activity against a variety of malignant neoplasms. It is also used as an inhibitor of thymidylate synthetase and DNA synthesis. In addition to this, it plays an important role in the treatment of colorectal cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol, isopropyl alcohol, methanol, aqueous buffers, acetone, dimethyl sulfoxide, and ethanol. Insoluble in ether, benzene and chloroform.

Notes
Store in a cool place.Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-91-9
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00006530
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
PubChem CID 5790
ChEBI CHEBI:60761
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 148°C to 151°C
Quantity 100 mg
UN Number UN2811
Beilstein 2645818
Merck Index 14,4112
Solubility Information Soluble in water,alcohol,isopropyl alcohol,methanol,aqueous buffers,acetone,dimethyl sulfoxide,and ethanol. Insoluble in ether,benzene and chloroform.
Formula Weight 246.2
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2'-deoxyuridine
compliance-icons compliance-icons
Product Identifier
  • 5-Fluoro-2'-deoxyuridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Floxuridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.