missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2,4-dichloropyrimidine, TRC

Product Code. 30399643
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399643 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399643 Supplier Toronto Research Chemicals Supplier No. TRCF5909601G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluoro-2,4-dichloropyrimidine
CAS 2927-71-1
Molecular Formula C4 H Cl2 F N2
Quantity 1 g
Synonym Pyrimidine, 2,4-dichloro-5-fluoro- (7CI, 8CI, 9CI, ACI), 2,4-Dichloro-5-fluoropyrimidine (ACI), 2,6-Dichloro-5-fluoropyrimidine, 5-Fluoro-2,4-dichloropyrimidine, NSC 70442
InChI Formula InChI=1S/C4HCl2FN2/c5-3-2(7)1-8-4(6)9-3/h1H
SMILES Fc1cnc(Cl)nc1Cl
IUPAC Name 2,4-dichloro-5-fluoropyrimidine
Molecular Weight (g/mol) 166.97
Formula Weight 165.9501
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.