missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2,4-dichloropyrimidine, 98%

Product Code. 11356789
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356789 1 g 1g
11366789 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356789 Supplier Thermo Scientific Alfa Aesar Supplier No. H26288.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2927-71-1 | C4HCl2FN2 | 166.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2927-71-1
Molecular Formula C4HCl2FN2
Molecular Weight (g/mol) 166.96
MDL Number MFCD00233551
InChI Key WHPFEQUEHBULBW-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine
PubChem CID 250705
IUPAC Name 2,4-dichloro-5-fluoropyrimidine
SMILES FC1=CN=C(Cl)N=C1Cl

Specifications

Melting Point 36°C to 39°C
Boiling Point 80°C (16 mmHg)
Flash Point 106°C (223°F)
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 166.97
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2,4-dichloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.