missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-1-indanone, 98%, Thermo Scientific™

Product Code. 10167453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167453 1 g Glass bottle 1g
10571121 250 mg Glass bottle 250mg
10434535 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10167453 Supplier Thermo Scientific Acros Supplier No. 271110010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-84-5
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.15
MDL Number MFCD00041031
InChI Key WVPPBVAMKNQXJA-UHFFFAOYSA-N
Synonym 5-fluoro-1-indanone, 5-fluoro-2,3-dihydro-1h-inden-1-one, 5-fluoroindan-1-one, 1h-inden-1-one, 5-fluoro-2,3-dihydro, 2,3-dihydro-5-fluoro-1h-inden-1-one, 5-fluoroindanone, 5-fluoro indanone, pubchem2388, 5-flouro-1-indanone, pubchem13785
PubChem CID 136537
SMILES FC1=CC=C2C(=O)CCC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 35°C to 39°C
Color White to Yellow
Boiling Point 92°C to 93°C (3.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
IUPAC Name 5-fluoro-2,3-dihydro-1H-inden-1-one
Formula Weight 150.15
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 5-Fluoro-1-indanone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Fluoro-1-indanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.