missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-1-indanone, 98%, Thermo Scientific™

Product Code. p-134731 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10571121

Brand: Thermo Scientific Acros 271112500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-84-5
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.15
MDL Number MFCD00041031
InChI Key WVPPBVAMKNQXJA-UHFFFAOYSA-N
Synonym 5-fluoro-1-indanone, 5-fluoro-2,3-dihydro-1h-inden-1-one, 5-fluoroindan-1-one, 1h-inden-1-one, 5-fluoro-2,3-dihydro, 2,3-dihydro-5-fluoro-1h-inden-1-one, 5-fluoroindanone, 5-fluoro indanone, pubchem2388, 5-flouro-1-indanone, pubchem13785
PubChem CID 136537
SMILES FC1=CC=C2C(=O)CCC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 35°C to 39°C
Color White to Yellow
Boiling Point 92°C to 93°C (3.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble
IUPAC Name 5-fluoro-2,3-dihydro-1H-inden-1-one
Formula Weight 150.15
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 5-Fluoro-1-indanone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Fluoro-1-indanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.