missing translation for 'onlineSavingsMsg'
Learn More

5-Ethynyluridine, TRC

Product Code. 30382238
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382238 10 mg
30381677 50 mg
30373485 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382238 Supplier Toronto Research Chemicals Supplier No. TRCE93265010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69075-42-9
Molecular Formula C11H12N2O6
Molecular Weight (g/mol) 268.22
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(C#C)C(=O)NC2=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H12N2O6/c1-2-5-3-13(11(18)12-9(5)17)10-8(16)7(15)6(4-14)19-10/h1,3,6-8,10,14-16H,4H2,(H,12,17,18)/t6-,7-,8-,10-/m1/s1
Formula Weight 268.0695
Chemical Name or Material 5-Ethynyluridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.