missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5'-Ethynyl-2'-deoxyuridine

Product Code. 17291222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17291222 1 g 1g
17201232 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17291222 Supplier Thermo Scientific Chemicals Supplier No. 466380010
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

252.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 252.23
Product Type 5 '-Ethynyl-2 '-deoxyuridine
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol
compliance-icons
Product Identifier
  • 5'-Ethynyl-2'-deoxyuridine
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H340-May cause genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5'-Ethynyl-2'-deoxyuridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.