Learn More
5-Ethylidene-2-norbornene, endo + exo, 98%, stab. with 100ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Description
5-Ethylidene-2-norbornene is used as a self healing agent for polymeric composites. It is also used to reinforce multiwalled carbon nanotubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.89 g/mL |
| Boiling Point | 146°C |
| Flash Point | 38°C (101°F) |
| Quantity | 500 mL |
| UN Number | UN3295 |
| Solubility Information | Soluble in water (0.01% at 20C). |
| Formula Weight | 120.19 |
| Percent Purity | 98% |
| Physical Form | Liquid |
| Chemical Name or Material | 5-Ethylidene-2-norbornene, endo + exo, Stabilized with 100ppm 4-tert-butylcatechol |
Safety and Handling
- 5-Ethylidene-2-norbornene
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Ethylidene norbornene
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.