missing translation for 'onlineSavingsMsg'
Learn More

5-Ethylidene-2-norbornene, endo + exo, 98%, stab. with 100ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Product Code. 11345028
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345028 100 mL 100mL
11335028 25 mL 25mL
11480640 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345028 Supplier Thermo Scientific Alfa Aesar Supplier No. 043972.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5-Ethylidene-2-norbornene is used as a self healing agent for polymeric composites. It is also used to reinforce multiwalled carbon nanotubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16219-75-3
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00167576
InChI Key OJOWICOBYCXEKR-WHZSQGQPSA-N
Synonym 5-ethylidene-2-norbornene, unii-85015b2v0r component, unii-q4d40uvr0r component
PubChem CID 12456033
IUPAC Name (1R,4R,5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
SMILES CC=C1CC2CC1C=C2

Specifications

Density 0.89 g/mL
Boiling Point 146°C
Flash Point 38°C (101°F)
Quantity 100 mL
UN Number UN3295
Solubility Information Soluble in water (0.01% at 20C).
Formula Weight 120.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Ethylidene-2-norbornene, endo + exo, Stabilized with 100ppm 4-tert-butylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.