missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Ethyl-2-pyridineethanol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 5-Ethyl-2-pyridine Ethanol |
| CAS | 5223-06-3 |
| Molecular Formula | C9 H13 N O |
| Quantity | 1 g |
| Synonym | 2-Pyridineethanol, 5-ethyl-, 5-Ethyl-2-pyridineethanol, 2-(5-Ethyl-2-pyridinyl)ethanol, 2-(5-Ethyl-2-pyridyl)ethanol, 2-(5-Ethylpyridin-2-yl)ethanol, 5-Ethyl-2-(2-hydroxyethyl)pyridine, 5-Ethyl-2-(β-hydroxyethyl)pyridine |
| InChI Formula | InChI=1S/C9H13NO/c1-2-8-3-4-9(5-6-11)10-7-8/h3-4,7,11H,2,5-6H2,1H3 |
| SMILES | CCc1ccc(CCO)nc1 |
| IUPAC Name | 2-(5-ethylpyridin-2-yl)ethanol |
| Molecular Weight (g/mol) | 151.21 |
| Formula Weight | 151.0997 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction