missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-methylpyridine, 97%, Thermo Scientific Chemicals

Product Code. 11317046
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317046 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11317046 Supplier Thermo Scientific Alfa Aesar Supplier No. L06901.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-90-5 | Molecular Formula: C8H11N | Molecular Weight: 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-90-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00006344
InChI Key NTSLROIKFLNUIJ-UHFFFAOYSA-N
Synonym aldehydine, 5-ethyl-2-picoline, 2-methyl-5-ethylpyridine, aldehydkollidin, aldehydecollidine, pyridine, 5-ethyl-2-methyl, 2,5-aldehydine, 2-picoline, 5-ethyl, 3-ethyl-6-methylpyridine, 6-methyl-3-ethylpyridine
PubChem CID 7728
IUPAC Name 5-ethyl-2-methylpyridine
SMILES CCC1=CC=C(C)N=C1

Specifications

Melting Point -71°C
Density 0.919
Boiling Point 178°C to 179°C
Flash Point 66°C (150°F)
Refractive Index 1.497
Quantity 100 g
UN Number UN2300
Beilstein 109269
Merck Index 14,3844
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material 5-Ethyl-2-methylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.