missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-1-nonanol, TRC

Product Code. 30381953
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381953 50 mg 50mg
30373254 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381953 Supplier Toronto Research Chemicals Supplier No. TRCE42002550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Ethyl-1-nonanol
CAS 137008-30-1
Molecular Formula C11 H24 O
Quantity 50 mg
Synonym 5-Ethyl-1-nonanol
InChI Formula InChI=1S/C11H24O/c1-3-5-8-11(4-2)9-6-7-10-12/h11-12H,3-10H2,1-2H3
SMILES CCCCC(CC)CCCCO
IUPAC Name 5-ethylnonan-1-ol
Molecular Weight (g/mol) 172.308
Formula Weight 172.183
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.