missing translation for 'onlineSavingsMsg'
Learn More

5-(Ethoxycarbonyl)thiophene-3-boronic Acid, TRC

Product Code. 30385666
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385666 250 mg
30381324 500 mg
30381112 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385666 Supplier Toronto Research Chemicals Supplier No. TRCE895058250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957121-19-6
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.02
IUPAC Name (5-ethoxycarbonylthiophen-3-yl)boronic acid
SMILES CCOC(=O)c1cc(cs1)B(O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H9BO4S/c1-2-12-7(9)6-3-5(4-13-6)8(10)11/h3-4,10-11H,2H2,1H3
Formula Weight 200.0315
Chemical Name or Material 5-(Ethoxycarbonyl)thiophene-3-boronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.