missing translation for 'onlineSavingsMsg'
Learn More

5-DOXYL-stearic Acid, Free Radical (>90%), TRC

Product Code. 30005718
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005718 5 mg 5mg
30011392 10 mg 10mg
30004892 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005718 Supplier Toronto Research Chemicals Supplier No. TRCD5355285MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29545-48-0
Molecular Formula C22H42NO4
Molecular Weight (g/mol) 384.57
Synonym 2-(3-Carboxypropyl)-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxy, 2-(3-Carboxypropyl)-2-tridecyl-4,4-dimethyl oxazoline-3-oxyl, 2-(3-Carboxypropyl)-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxyl, 5-DSA, 5-Doxylstearate, 5-Doxylstearic acid, 5-Nitroxystearate, 5-SASL, 5NS, K 5D, K 5D (spin probe), 2-(3--Carboxypropyl)-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxy
SMILES OS(C(F)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)F)(=O)=O

Specifications

Quantity 5 mg
Percent Purity >95%
Chemical Name or Material 5-DOXYL-stearic Acid Free Radical
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.