missing translation for 'onlineSavingsMsg'
Learn More

5-(Diisopropylamino)amylamine, Thermo Scientific™

Product Code. 10321330
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10321330 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321330 Supplier Thermo Scientific Maybridge Supplier No. MAY00263DE
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 209803-40-7
Packaging Amber Glass Bottle
Molecular Formula C11H26N2
Quantity 5 g
Synonym 5-diisopropylamino amylamine, n,n-diisopropyl-1,5-pentanediamine, 5-aminopentyl diisopropylamine, n1,n1-diisopropylpentane-1,5-diamine, 5-aminopentyl bis methylethyl amine, acmc-1cpdx, n1,n1-dsopropylpentane-1,5-damne, n',n'-di propan-2-yl pentane-1,5-diamine, 1,5-pentanediamine,n1,n1-bis 1-methylethyl
InChI Key CIFBCEXYFFKMEB-UHFFFAOYSA-N
SMILES CC(C)N(CCCCCN)C(C)C
IUPAC Name N',N'-di(propan-2-yl)pentane-1,5-diamine
Molecular Weight (g/mol) 186.343
PubChem CID 3790585
Formula Weight 186.34
Percent Purity 97%
Chemical Name or Material 5-(Diisopropylamino)amylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.