missing translation for 'onlineSavingsMsg'
Learn More

5-Cyclopropyl-1H-pyrazole-3-carboxylic acid, 94%, Thermo Scientific™

Product Code. 11479460
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11479460 250 mg 250mg
11489460 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11479460

Brand: Thermo Scientific Alfa Aesar H50306.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid is used as organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401629-04-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD04967194
InChI Key GNWMHLBUCXEXPE-UHFFFAOYSA-N
Synonym 3-cyclopropyl-1h-pyrazole-5-carboxylic acid, 3-cyclopropylpyrazole-5-carboxylic acid, 5-cyclopropyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-cyclopropyl, 5-cyclopropylpyrazole-3-carboxylic acid, acmc-1ap8l, 1h-pyrazole-3-carboxylic acid, 5-cyclopropyl, 1h-pyrazole-3-carboxylic acid, 5-cyclopropyl-9ci
PubChem CID 776464
IUPAC Name 5-cyclopropyl-1H-pyrazole-3-carboxylic acid
SMILES C1CC1C2=CC(=NN2)C(=O)O

Specifications

Melting Point ∽175°C (decomposition)
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 152.15
Percent Purity 94%
Chemical Name or Material 5-Cyclopropyl-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.