missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-thiophene carbaldehyde, Thermo Scientific™

Product Code. 10381420
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10381420 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10381420 Supplier Thermo Scientific Maybridge Supplier No. MAY00111DA
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 21512-16-3
Packaging Amber Glass Bottle
Molecular Formula C6H3NOS
Quantity 1 g
Synonym 5-cyano-2-thiophene carbaldehyde, 5-cyano-2-thiophenecarboxaldehyde, 5-cyanothiophene-2-carboxaldehyde, 2-thiophenecarbonitrile, 5-formyl, 5-cyano-2-thiophenecarbaldehyde, 2-formyl-5-cyanothiophen, 2-cyano-5-formylthiophene, 2-formyl-5-cyanothiophene, 2-cyano-5-formyl-thiophene, 5-cyanothiophene-2-aldehyde
InChI Key PZIFYWVUYHMYOA-UHFFFAOYSA-N
SMILES C1=C(SC(=C1)C#N)C=O
IUPAC Name 5-formylthiophene-2-carbonitrile
Molecular Weight (g/mol) 137.156
PubChem CID 12280004
Formula Weight 137.16
Percent Purity 97%
Chemical Name or Material 5-Cyano-2-thiophene carbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.