missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-methyl-4-phenylthiazole, 97%, Thermo Scientific™

Catalog Number 15464388
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
84302-57-8
C11H8N2S
200.259
MFCD22682923
CLGMFVPFRMXIRG-UHFFFAOYSA-N
5-cyano-2-methyl-4-phenylthiazole, 2-methyl-4-phenyl-5-thiazolecarbonitrile
12793598
2-methyl-4-phenyl-1,3-thiazole-5-carbonitrile
CC1=NC(=C(S1)C#N)C2=CC=CC=C2
This item is not returnable. View return policy
84302-57-8
C11H8N2S
200.259
MFCD22682923
CLGMFVPFRMXIRG-UHFFFAOYSA-N
5-cyano-2-methyl-4-phenylthiazole, 2-methyl-4-phenyl-5-thiazolecarbonitrile
12793598
2-methyl-4-phenyl-1,3-thiazole-5-carbonitrile
CC1=NC(=C(S1)C#N)C2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Beilstein 5933758
Formula Weight 200.26
Percent Purity 97%
Chemical Name or Material 5-Cyano-2-methyl-4-phenylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'