missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cyano-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
£69.82 - £221.00
Chemical Identifiers
CAS | 863868-29-5 |
---|---|
Molecular Formula | C13H15BFNO2 |
Molecular Weight (g/mol) | 247.08 |
MDL Number | MFCD07437899 |
InChI Key | DCBKFUVWBRFDDD-UHFFFAOYSA-N |
Synonym | 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 5-cyano-2-fluorophenylboronic acid, pinacol ester, 5-cyano-2-fluorobenzeneboronic acid pinacol ester, 5-cyano-2-fluorophenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
PubChem CID | 53426612 |
IUPAC Name | 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile |
SMILES | CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C#N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
863868-29-5 | |
247.08 | |
DCBKFUVWBRFDDD-UHFFFAOYSA-N | |
53426612 | |
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C#N |
C13H15BFNO2 | |
MFCD07437899 | |
4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 5-cyano-2-fluorophenylboronic acid, pinacol ester, 5-cyano-2-fluorobenzeneboronic acid pinacol ester, 5-cyano-2-fluorophenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile |
Specifications
863868-29-5 | |
MFCD07437899 | |
DCBKFUVWBRFDDD-UHFFFAOYSA-N | |
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile | |
53426612 | |
96% | |
5-Cyano-2-fluorobenzeneboronic acid pinacol ester |
C13H15BFNO2 | |
4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 5-cyano-2-fluorophenylboronic acid, pinacol ester, 5-cyano-2-fluorobenzeneboronic acid pinacol ester, 5-cyano-2-fluorophenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 4-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C#N | |
247.08 | |
247.07 | |
Solid |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures