missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-(3-methyl-2-pyridyl)-4-phenylthiazole, 97%, Thermo Scientific™

Catalog Number 15494106
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1858241-97-0
C16H11N3S
277.345
MFCD22682957
GNCHRPZBZFYZAD-UHFFFAOYSA-N
2-3-methylpyridin-2-yl-4-phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-2-3-methyl-2-pyridyl-4-phenylthiazole
98000062
2-(3-methylpyridin-2-yl)-4-phenyl-1,3-thiazole-5-carbonitrile
CC1=C(N=CC=C1)C2=NC(=C(S2)C#N)C3=CC=CC=C3
This item is not returnable. View return policy
1858241-97-0
C16H11N3S
277.345
MFCD22682957
GNCHRPZBZFYZAD-UHFFFAOYSA-N
2-3-methylpyridin-2-yl-4-phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-2-3-methyl-2-pyridyl-4-phenylthiazole
98000062
2-(3-methylpyridin-2-yl)-4-phenyl-1,3-thiazole-5-carbonitrile
CC1=C(N=CC=C1)C2=NC(=C(S2)C#N)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 277.34
Percent Purity 97%
Chemical Name or Material 5-Cyano-2-(3-methyl-2-pyridyl)-4-phenylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'