missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovanillic acid, tech. 90%

Product Code. p-7020774
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11495233

Brand: Thermo Scientific Alfa Aesar A12841.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62936-23-6 | C8H7ClO4 | 202.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62936-23-6
Molecular Formula C8H7ClO4
Molecular Weight (g/mol) 202.59
MDL Number MFCD00016531
InChI Key XBRYEHVBBMSSCG-UHFFFAOYSA-N
PubChem CID 44215
IUPAC Name 3-chloro-4-hydroxy-5-methoxybenzoic acid
SMILES COC1=C(C(=CC(=C1)C(=O)O)Cl)O

Specifications

Melting Point 242°C to 248°C
Quantity 5 g
Beilstein 2648897
Synonym 5-chlorovanillic acid, 3-chloro-4-hydroxy-5-methoxy-benzoic acid, benzoic acid, 3-chloro-4-hydroxy-5-methoxy, 3-chloro-4-hydroxy-5-methoxy benzoic acid, 5-chlorovanillicacid, 5-chloro-vanillic acid, acmc-1b2hg, chloro-4-hydroxy-3-methoxybenzoic acid, 5-chloro-4-hydroxy-3-methoxybenzoic acid, benzoic acid,3-chloro-4-hydroxy-5-methoxy
Formula Weight 202.6
Percent Purity 90%
Grade Technical
Chemical Name or Material 5-Chlorovanillic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.