missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleroyl chloride, 98%

Product Code. p-7037489
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 15422688

Brand: Thermo Scientific Alfa Aesar H56237.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1575-61-7 | C5H8Cl2O | 155.02 g/mol

5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorovaleroyl chloride (5-CVC) has been used in the development and validation of a GC-FID method for the analysis of low level impurities present in 5-CVC. It has been used in the synthesis of anion-exchange resins by acylation of polystyrene/divinylbenzene and subsequent amination with trimethylamine. It is commonly used as alkylating agent in the synthesis of pharmaceutical intermediates and other specialty chemicals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, bases, alcohols, amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1575-61-7
Molecular Formula C5H8Cl2O
Molecular Weight (g/mol) 155.02
MDL Number MFCD00000758
InChI Key SVNNWKWHLOJLOK-UHFFFAOYSA-N
Synonym 5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1
PubChem CID 74089
IUPAC Name 5-chloropentanoyl chloride
SMILES ClCCCCC(Cl)=O

Specifications

Density 1.2
Boiling Point 38°C to 39°C
Flash Point 91°C (196°F)
Linear Formula Cl(CH2)4COCl
Refractive Index 1.464
Quantity 50 g
UN Number UN3265
Beilstein 1745182
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 155.02
Percent Purity 98%
Chemical Name or Material 5-Chlorovaleroyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.