missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorovaleronitrile, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 5-Chlorovaleronitrile |
| CAS | 6280-87-1 |
| Molecular Formula | C5H8ClN |
| Quantity | 5 g |
| Synonym | 1-Chloro-4-cyanobutane, 4-Chloro-1-cyanobutane, 5-Chloropentanenitrile, NSC 6156, δ-Chlorovaleronitrile |
| InChI Formula | InChI=1S/C5H8ClN/c6-4-2-1-3-5-7/h1-4H2 |
| SMILES | ClCCCCC#N |
| IUPAC Name | 5-chloropentanenitrile |
| Molecular Weight (g/mol) | 117.58 |
| Formula Weight | 117.03 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction