missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleronitrile, 97%, Thermo Scientific™

Product Code. 10336030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336030 25 mL Glass bottle 25mL
10714411 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336030 Supplier Thermo Scientific Supplier No. 110140250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6280-87-1
Molecular Formula C5H8ClN
Molecular Weight (g/mol) 117.58
MDL Number MFCD00001977
InChI Key JSAWFGSXRPCFSW-UHFFFAOYSA-N
Synonym 5-chlorovaleronitrile, pentanenitrile, 5-chloro, 5-chloro-n-valeronitrile, valeronitrile, 5-chloro, 5-chloro valeronitrile, .delta.-chlorovaleronitrile, snxhfhaiuzjp@, 5-chlorovaieronitrile, 4-chlorobutyl cyanide, delta-chlorovaleronitrile
PubChem CID 80474
IUPAC Name 5-chloropentanenitrile
SMILES ClCCCCC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.0400g/mL
Boiling Point 92°C to 93°C (10.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)4CN
Refractive Index 1.4452 to 1.4472
Quantity 25 mL
Specific Gravity 1.04
Formula Weight 117.58
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Chlorovaleronitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chlorovaleronitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.