Learn More
5-Chlorothiophene-2-sulfonyl chloride, 97%
Description
5-Chlorothiophene-2-sulfonyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs. It is used in the preparation of Notch-1-sparing γ-secretase inhibitors for the treatment of Alzheimer's disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Not miscible in water.
Notes
Moisture Sensitive. Store at 4°C. Incomaptible with oxidizing agents and bases.
Specifications
Specifications
| Density | 1.623 |
| Boiling Point | 117°C to 120°C (14 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.586 |
| Quantity | 10 g |
| UN Number | UN3265 |
| Beilstein | 130710 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Not miscible in water. |
| Formula Weight | 217.09 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.