missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-carboxylic acid, 98%

Product Code. 11412228
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412228 5 g 5g
11422228 25 g 25g
11432228 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412228 Supplier Thermo Scientific Alfa Aesar Supplier No. A15209.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24065-33-6 | C5H3ClO2S | 162.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24065-33-6
Molecular Formula C5H3ClO2S
Molecular Weight (g/mol) 162.59
MDL Number MFCD00041426
InChI Key QZLSBOVWPHXCLT-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-chloro, 2-chloro-5-carboxythiophene, 5-chloro-2-thiophene carboxylic acid, pubchem7380, maybridge3_003717, acmc-209g8i, rivaroxaban related compound, 5-18-06-00177 beilstein handbook reference, ksc204m2j
PubChem CID 95048
IUPAC Name 5-chlorothiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 149°C to 152°C
Quantity 5 g
Beilstein 118361
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 5-Chlorothiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.