missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorothiophene-2-carboxaldehyde, 97%

Product Code. 11449867
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449867 1 g 1g
11459867 5 g 5g
11469867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449867 Supplier Thermo Scientific Alfa Aesar Supplier No. B22833.03
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 7283-96-7 | C5H3ClOS | 146.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7283-96-7
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00047090
InChI Key VWYFITBWBRVBSW-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxaldehyde, 2-chloro-5-thiophenecarboxaldehyde, 5-chlorothiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 5-chloro, 5-chloro-2-formylthiophene, 2-chloro-5-formylthiophene, 5-chlorothiophene-2-aldehyde, 5-chloro-2-thiophenecarbaldehyde, 5-chloro-2-thiophene-carboxaldehyde, pubchem7742
PubChem CID 81700
IUPAC Name 5-chlorothiophene-2-carbaldehyde
SMILES ClC1=CC=C(S1)C=O

Specifications

Density 1.376
Boiling Point 88°C to 90°C (9 mmHg)
Flash Point 97°C (208°F)
Refractive Index 1.604
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 46.6
Percent Purity 97%
Chemical Name or Material 5-Chlorothiophene-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.