missing translation for 'onlineSavingsMsg'
Learn More

5-chlorosulfonyl-2-hydroxybenzoic acid, 97%, Thermo Scientific™

Product Code. 10324622
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10324622

Brand: Thermo Scientific Maybridge CD11776DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17243-13-9
Molecular Formula C7H5ClO5S
Molecular Weight (g/mol) 236.62
MDL Number MFCD00052974
InChI Key SJXHSFSHNKFRLN-UHFFFAOYSA-N
Synonym 5-chlorosulfonyl-2-hydroxybenzoic acid, 5-chlorosulfonyl-2-hydroxy-benzoic acid, 5-chlorosulfonyl salicylic acid, benzoic acid, 5-chlorosulfonyl-2-hydroxy, benzoic acid,5-chlorosulfonyl-2-hydroxy, 5-chlorosulphonyl-2-hydroxybenzoic acid, 5-chlorosulfonyl-2-hydroxybenzoicacid, acmc-1cis5, 5-chlorosulphonyl salicylic acid
PubChem CID 2777240
IUPAC Name 5-chlorosulfonyl-2-hydroxybenzoic acid
SMILES OC(=O)C1=C(O)C=CC(=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 236.63
Percent Purity 97%
Chemical Name or Material 5-Chlorosulfonyl-2-hydroxybenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.