missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosulfonyl-2-fluorobenzoic acid, 97%

Product Code. p-7032091
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15496237

Brand: Thermo Scientific Alfa Aesar H33431.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 37098-75-2 | C7H4ClFO4S | 238.613 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37098-75-2
Molecular Formula C7H4ClFO4S
Molecular Weight (g/mol) 238.613
MDL Number MFCD03030335
InChI Key CISMZCVQNVNWJW-UHFFFAOYSA-N
Synonym 5-chlorosulfonyl-2-fluorobenzoic acid, 2-fluoro-5-chlorosulfonyl benzoic acid, 5-chlorosulphonyl-2-fluorobenzoic acid, 5-chlorosulfonyl-2-fluoro-benzoic acid, benzoic acid, 5-chlorosulfonyl-2-fluoro, 5-chlorosulfonyl-2-fluorobenzoicacid, acmc-1afaw, ksc577o0r, 2-fluoro-5-chlorosulfonylbenzoic acid
PubChem CID 608984
IUPAC Name 5-chlorosulfonyl-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(=O)O)F

Specifications

Melting Point 111°C to 113°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 238.62
Percent Purity 97%
Chemical Name or Material 5-Chlorosulfonyl-2-fluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.