missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylic Acid, TRC

Product Code. 30360678
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360678 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360678 Supplier Toronto Research Chemicals Supplier No. TRCC38021550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chlorosalicylic Acid
CAS 321-14-2
Molecular Formula C7H5ClO3
Quantity 50 g
Synonym 5-Chloro-2-hydroxybenzoic Acid, NSC 30111, 2-Hydroxy-5-chlorobenzoic Acid
InChI Formula InChI=1S/C7H5ClO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,(H,10,11)
SMILES OC(=O)c1cc(Cl)ccc1O
IUPAC Name 5-chloro-2-hydroxybenzoic acid
Molecular Weight (g/mol) 172.57
Formula Weight 171.9927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.