missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylaldehyde, 98%

Product Code. 10116990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10116990 25 g Plastic bottle 25g
10580934 5 g Glass bottle 5g
10471011 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10116990 Supplier Thermo Scientific Acros Supplier No. 405030250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 635-93-8 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-93-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00003331
InChI Key FUGKCSRLAQKUHG-UHFFFAOYSA-N
Synonym 5-chlorosalicylaldehyde, 2-hydroxy-5-chlorobenzaldehyde, benzaldehyde, 5-chloro-2-hydroxy, 4-chloro-2-formylphenol, salicylaldehyde, 5-chloro, 5-chloro-2-hydroxy-benzaldehyde, unii-4gcc8zkm3o, 5-chloro-salicylaldehyde, 4gcc8zkm3o, 5-chloro-2-hydroxy benzaldehyde
PubChem CID 12481
IUPAC Name 5-chloro-2-hydroxybenzaldehyde
SMILES OC1=CC=C(Cl)C=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 102°C to 103°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 156.57
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorosalicylaldehyde, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.