missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroquinoline, Thermo Scientific™

Product Code. 17750669 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17750669

Brand: Thermo Scientific Acros 469740050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Alpha Vector CHLOROQUINOLINE
Chemical Name or Material 5-Chloroquinoline
CAS 635-27-8
Melting Point 28°C to 32°C
Flash Point 263°C
Packaging Glass bottle
Molecular Formula C9 H6 Cl N
MDL Number 00239413
Quantity 5 g
Molecular Weight (g/mol) 163.61
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.