missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloroquinolin-8-ol, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
5-Chloroquinolin-8-ol
Specifications
Specifications
| Chemical Name or Material | Cloxiquine |
| CAS | 130-16-5 |
| Analyte or Component Names | Cloxiquine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H6 Cl N O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cloxiquine, 5-Chloro-8-hydroxyquinoline, 5-Chloro-8-oxyquinoline, 5-Chloro-8-quinolinol, 5-Chloroquinoline-8-ol, 5-Chlorooxine, 8-Hydroxy-5-chloroquinoline, Cloxyquin, Dermofongin, Dermofungin, NSC 35083, NSC 53182, NSC 74943, Clioquinol Imp. A (EP), Clioquinol Impurity A |
| InChI Formula | InChI=1S/C9H6ClNO/c10-7-3-4-8(12)9-6(7)2-1-5-11-9/h1-5,12H |
| SMILES | Oc1ccc(Cl)c2cccnc12 |
| IUPAC Name | 5-chloroquinolin-8-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction