missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyrimidine, 95%

Product Code. 15589493 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15589493 1 g 1g
15599493 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15589493 Supplier Thermo Scientific Acros Supplier No. 459660010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17180-94-8 | C4H3ClN2 | 114.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17180-94-8
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD03840717
InChI Key ZLNPDTOTEVIMMY-UHFFFAOYSA-N
Synonym pyrimidine, 5-chloro, 5-chloro-pyrimidine, 5-choloropyrimidine, 5-chloranylpyrimidine, pubchem7041, pyrimidine, 5-chloro-6ci,8ci,9ci
PubChem CID 12600384
IUPAC Name 5-chloropyrimidine
SMILES ClC1=CN=CN=C1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 114.53
Percent Purity 95%
Chemical Name or Material 5-Chloropyrimidine
compliance-icons
Product Identifier
  • 5-Chloropyrimidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.