missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Chloropyridine-3-boronic acid, 95%, Thermo Scientific™

Artikelnummer. 11322919
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11322919 1 g 1g
11332919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11322919 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H27756.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 872041-85-5
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD06798243
InChI Key NJXYBTMCTZAUEE-UHFFFAOYSA-N
Synonym 5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397
PubChem CID 20111877
IUPAC Name (5-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC(Cl)=CN=C1

Tekniske data

Quantity 1 g
Formula Weight 157.37
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-3-boronic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.