missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15414007
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414007 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414007 Supplier Thermo Scientific Alfa Aesar Supplier No. H30923.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86873-60-1 | Molecular Formula: C6H4ClNO2 | Molecular Weight: 157.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86873-60-1
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD04114192
InChI Key GJLOKYIYZIOIPN-UHFFFAOYSA-N
Synonym 5-chloropicolinic acid, 5-chloro-2-picolinic acid, 5-chloro-2-pyridinecarboxylic acid, 5-chloro-pyridine-2-carboxylic acid, 5-chloro-2-carboxypyridine, 2-carboxy-5-chloropyridine, 5-chloropyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 5-chloro, pubchem10379, 5-chloro-picolinic acid
PubChem CID 2762872
IUPAC Name 5-chloropyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1Cl)C(=O)O

Specifications

Melting Point 166°C to 171°C
Quantity 1 g
Formula Weight 157.56
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.