missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridin-3-ol, 97%, Thermo Scientific™

Product Code. 10748253
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10748253 50 g 50g
10147112 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748253 Supplier Thermo Scientific Maybridge Supplier No. SB00891EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 74115-12-1
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00006379
InChI Key TUIDQYRZDZRHPQ-UHFFFAOYSA-N
Synonym 5-chloro-3-hydroxypyridine, 5-chloro-3-pyridinol, 3-chloro-5-hydroxypyridine, 3-pyridinol, 5-chloro, 5chloro-3-pyridinol, pubchem6628, 3-chloro-5-pyridinol, acmc-209otf, 5-chloranylpyridin-3-ol, 3chloro-5-hydroxypyridine
PubChem CID 582305
IUPAC Name 5-chloropyridin-3-ol
SMILES C1=C(C=NC=C1Cl)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula Cl(C5H3N)OH
Quantity 50 g
Formula Weight 129.55
Percent Purity 97%
Chemical Name or Material 5-Chloropyridin-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.