missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorooxindole, 98%

Product Code. 11449486
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11449486

Brand: Thermo Scientific Alfa Aesar A19210.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17630-75-0 | C8H6ClNO | 167.59 g/mol

5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system. 5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system. 5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate.

Solubility
Slightly Soluble (2.0 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17630-75-0
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00191927
InChI Key WWJLCYHYLZZXBE-UHFFFAOYSA-N
Synonym 5-chlorooxindole, 5-chloroindolin-2-one, 5-chloro-1,3-dihydro-2h-indol-2-one, 5-chloro-2-indolinone, 5-chloro-1,3-dihydro-indol-2-one, 5-chloro-2-oxindole, 2h-indol-2-one, 5-chloro-1,3-dihydro, 5-chloro-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-chloro, 5-chlorooxindol
PubChem CID 152801
SMILES ClC1=CC=C2NC(=O)CC2=C1

Specifications

Melting Point 196°C to 200°C
Quantity 5 g
Solubility Information Slightly Soluble (2.0g/L) (25°C).
IUPAC Name 5-chloro-2,3-dihydro-1H-indol-2-one
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 5-Chlorooxindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.