missing translation for 'onlineSavingsMsg'
Learn More

5-Chloronaphthalene-2-sulfonic Acid, Potassium Salt, TRC

Product Code. 30358989
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358989 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358989 Supplier Toronto Research Chemicals Supplier No. TRCC3709251G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloronaphthalene-2-sulfonic Acid, Potassium Salt
CAS 1024267-23-9
Molecular Formula C10 H6 Cl O3 S . K
Quantity 1 g
InChI Formula InChI=1S/C10H7ClO3S.K/c11-10-3-1-2-7-6-8(15(12,13)14)4-5-9(7)10;/h1-6H,(H,12,13,14);/q;+1/p-1
SMILES [K+].[O-]S(=O)(=O)c1ccc2c(Cl)cccc2c1
IUPAC Name potassium;5-chloronaphthalene-2-sulfonate
Molecular Weight (g/mol) 280.77
Formula Weight 279.9363
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.