missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™

Product Code. 10141212 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10141212 1 g 1g
10573022 250 mg 250mg
10692495 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10141212 Supplier Thermo Scientific Maybridge Supplier No. CD02742DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57238-75-2
Molecular Formula C9H6Cl2N2O
Molecular Weight (g/mol) 229.06
MDL Number MFCD00119077
InChI Key BJVYSQGEJHKTBW-UHFFFAOYSA-N
Synonym 5-chloromethyl-3-4-chlorophenyl-1,2,4-oxadiazole, 5-chloromethyl-3-4-chloro-phenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole, 5-chloromethyl-3-4-chlorophenyl, 5-chloromethyl-3-4-chlorophenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-chloromethyl-3-4-chlorophenyl, zlchem 1100
PubChem CID 2735758
IUPAC Name 5-(chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
SMILES ClCC1=NC(=NO1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 95%
Formula Weight 229.07
Percent Purity 95%
Chemical Name or Material 5-(Chloromethyl)-3-(4-chlorophenyl)-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.